Ethyl-4-chloroacetoacetate 97%

Supplier: Ambeed
Danger

Synonyms: Ethyl 4-chloro-3-oxobutanoate

A369533-25G A369533-100G
77115-392EA 5.31 USD
77115-392 77572-102
Ethyl-4-chloroacetoacetate 97%
Ethyl-4-chloroacetoacetate

Precaución: for Research Use Only

Formula: C₆H₉ClO₃
MW: 164.59 g/mol
MDL Number: MFCD00000939
CAS Number: 638-07-3
UN: 2922
ADR: 8,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR