N1,N2-Dibenzyloxalamide 95%

Supplier: Ambeed
Danger

A369231-1G A369231-5G A369231-25G
77115-390EA 16.98 USD
77115-390 77594-096 77686-040
N1,N2-Dibenzyloxalamide 95%
N1,N2-Dibenzyloxalamide

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₂O₂
MW: 268.31 g/mol
MDL Number: MFCD00022012
CAS Number: 3551-78-8
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR