BMS-863233 hydrochloride ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
XL413 hydrochloride, (S)-8-Chloro-2-(pyrrolidin-2-yl)benzofuro[3,2-d]pyrimidin-4(3H)-one xhydrochloride
A368616-10MG
A368616-1MG
A368616-5MG
A368616-100MG
A368616-50MG
77115-384EA
43.5
USD
77115-384
77115-386
77115-388
77686-037
77686-038
BMS-863233 hydrochloride ≥98%
BMS-863233 hydrochloride
Precaución: for Research Use Only
Formula:
C₁₄H₁₃Cl₂N₃O₂ MW: 326.18 g/mol |
MDL Number:
MFCD28023577 CAS Number: 1169562-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...