3-(4-Pyridyl)-DL-alanine 97%

Supplier: Ambeed
Warning

Synonyms: H-DL-3-(4-Pyridyl)-Ala-OH

A368534-1G A368534-250MG
77115-380EA 80.34 USD
77115-380 77115-382
3-(4-Pyridyl)-DL-alanine 97%
3-(4-Pyridyl)-DL-alanine

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MDL Number: MFCD05228021
CAS Number: 1956-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR