4,5-Difluoro-2-methylbenzoic acid 98%

Supplier: Ambeed
Warning

A367323-100G A367323-10G A367323-1G A367323-250MG A367323-25G A367323-5G
77115-368EA 543.84 USD
77115-368 77115-370 77115-372 77115-374 77115-376 77115-378
4,5-Difluoro-2-methylbenzoic acid 98%
4,5-Difluoro-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
MDL Number: MFCD08062377
CAS Number: 183237-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR