4-(4,6-Dichloropyrimidin-2-yl)morpholine 98%
Supplier: Ambeed
|
Warning
|
A366937-10G
A366937-1G
A366937-250MG
A366937-25G
A366937-5G
77115-352EA
99.73
USD
77115-352
77115-354
77115-356
77115-358
77115-360
4-(4,6-Dichloropyrimidin-2-yl)morpholine 98%
4-(4,6-Dichloropyrimidin-2-yl)morpholine
Precaución: for Research Use Only
Formula:
C₈H₉Cl₂N₃O MW: 234.08 g/mol |
MDL Number:
MFCD05022359 CAS Number: 10397-13-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...