α-Bromo-4-chloro-3-fluorotoluene 98%

Supplier: Ambeed
Danger

A364549-10G A364549-1G A364549-25G A364549-5G
77115-326EA 37.08 USD
77115-326 77115-328 77115-330 77115-332
α-Bromo-4-chloro-3-fluorotoluene 98%
α-Bromo-4-chloro-3-fluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrClF
MDL Number: MFCD04115859
CAS Number: 206362-80-3
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR