Cloquintocet mexyl 95%

Supplier: Ambeed
Warning

Synonyms: Cloquintocet-mexyl, 2-Heptyl 2-(5-Chloro-8-quinolinyloxy)acetate, 2-Heptyl 2-[(5-Chloroquinolin-8-yl)oxy]acetate, 2-[(5-Chloroquinolin-8-yl)oxy]acetic Acid 2-Heptyl Ester

A361564-1G A361564-5G A361564-25G A361564-100G
77115-310EA 9.55 USD
77115-310 77115-312 77613-060 77686-023
Cloquintocet mexyl 95%
Cloquintocet mexyl

Precaución: for Research Use Only

Formula: C₁₈H₂₂ClNO₃
MW: 335.83 g/mol
MDL Number: MFCD01632329
CAS Number: 99607-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR