2-Chloro-4,6-dimethoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A360815-100MG A360815-1G A360815-250MG
77115-286EA 15.45 USD
77115-286 77115-288 77115-290
2-Chloro-4,6-dimethoxybenzaldehyde 98%
2-Chloro-4,6-dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MDL Number: MFCD05863686
CAS Number: 82477-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR