3-(Benzylthio)propanoic acid 98%

Supplier: Ambeed
Danger

Synonyms: Mpa(Bzl)

A358674-10G A358674-1G A358674-25G A358674-5G
77115-262EA 61.54 USD
77115-262 77115-264 77115-266 77115-268
3-(Benzylthio)propanoic acid 98%
3-(Benzylthio)propanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂S
MW: 196.27 g/mol
MDL Number: MFCD00020521
CAS Number: 2899-66-3
UN: 3335
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR