1,2-Bis(phenylsulfonyl)ethane 97+%

Supplier: Ambeed
Warning

A358665-1G A358665-250MG A358665-25G A358665-5G
77115-254EA 24.41 USD
77115-254 77115-256 77115-258 77115-260
1,2-Bis(phenylsulfonyl)ethane 97+%
1,2-Bis(phenylsulfonyl)ethane

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₄S₂
MW: 310.39 g/mol
MDL Number: MFCD00041256
CAS Number: 599-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR