1-Bromo-3,4-dichloro-5-fluorobenzene 98%

Supplier: Ambeed
Warning

A357674-1G A357674-250MG A357674-5G A357674-25G
77115-248EA 8.24 USD
77115-248 77115-250 77115-252 77568-728
1-Bromo-3,4-dichloro-5-fluorobenzene 98%
1-Bromo-3,4-dichloro-5-fluorobenzene

Precaución: For research use only.

Formula: C₆H₂BrCl₂F
MDL Number: MFCD09878164
CAS Number: 1000572-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR