3-Bromo-5-methylpyridine 96%

Supplier: Ambeed
Warning

A357562-100G A357562-10G A357562-25G A357562-500G A357562-5G
77115-236EA 159.65 USD
77115-236 77115-238 77115-240 77115-242 77115-244 77115-246
3-Bromo-5-methylpyridine 96%
3-Bromo-5-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₆BrN
MW: 172.02
MDL Number: MFCD01646141
CAS Number: 3430-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR