Tazarotene 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 6-((4,4-dimethylthiochroman-6-yl)ethynyl)nicotinate

A357060-100MG A357060-10MG A357060-1G A357060-1MG A357060-250MG A357060-25MG A357060-50MG
77115-220EA 103 USD
77115-220 77115-222 77115-224 77115-226 77115-228 77115-230 77115-232
Tazarotene 97%
Tazarotene

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₂S
MW: 351.46 g/mol
MDL Number: MFCD00867628
CAS Number: 118292-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR