p-Tolylhydrazinium chloride 95%

Supplier: Ambeed
Danger

Synonyms: 4-Methylphenylhydrazinium chloride, 4-Tolylhydrazine hydrochloride, p-Tolylhydrazine hydrochloride, 1-(4-Methylphenyl)hydrazine hydrochloride, 4-Methylphenylhydrazine hydrochloride, 1-(4-Tolyl)hydrazine hydrochloride

A356721-10G A356721-25G A356721-5G A356721-100G
77115-212EA 6.18 USD
77115-212 77115-214 77115-216 77686-015
p-Tolylhydrazinium chloride 95%
p-Tolylhydrazinium chloride

Precaución: for Research Use Only

Formula: C₇H₁₁ClN₂
MDL Number: MFCD00012940
CAS Number: 637-60-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR