EDTA disodium salt 98%
Supplier: Ambeed
|
Danger
|
A354454-1000G
A354454-100G
A354454-500G
77115-160EA
17.52
USD
77115-160
77115-162
77115-164
77115-166
EDTA disodium salt 98%
EDTA disodium salt
Precaución: for Research Use Only
Formula:
C₁₀H₁₄N₂Na₂O₈ |
MDL Number:
MFCD00070672 CAS Number: 139-33-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...