4-Ethyl-5-fluoro-6-hydroxypyrimidine 98%

Supplier: Ambeed
Warning

A352124-1G A352124-5G
77115-144EA 6.37 USD
77115-144 77686-001
4-Ethyl-5-fluoro-6-hydroxypyrimidine 98%
4-Ethyl-5-fluoro-6-hydroxypyrimidine

Precaución: for Research Use Only

Formula: C₆H₇FN₂O
MDL Number: MFCD08063343
CAS Number: 137234-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR