3,9-Bis(2,4-di-tert-butylphenoxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane ≥97% (by GC)
Supplier: Ambeed
|
Warning
|
Synonyms:
Pentaerythritol bis(2,4-di-tert-butylphenylphosphite)
A351864-1G
A351864-25G
A351864-5G
A351864-100G
A351864-500G
A351864-1000G
77115-138EA
5.31
USD
77115-138
77115-140
77115-142
77568-706
77594-050
77685-999
3,9-Bis(2,4-di-tert-butylphenoxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane ≥97% (by GC)
3,9-Bis(2,4-di-tert-butylphenoxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane
Precaución: For research use only.
Formula:
C₃₃H₅₀O₆P₂ MW: 604.70 g/mol |
MDL Number:
MFCD00071521 CAS Number: 26741-53-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...