Benzyl 3-hydroxypiperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A351272-1G A351272-25G A351272-5G
77115-114EA 9.26 USD
77115-114 77115-116 77115-118
Benzyl 3-hydroxypiperidine-1-carboxylate 98%
Benzyl 3-hydroxypiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₃
MW: 235.27 g/mol
MDL Number: MFCD03839903
CAS Number: 95798-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR