5-Bromo-3-chloro-2-methoxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxy-3-chloro-5-bromopyridine

A348354-1G A348354-250MG A348354-25G A348354-5G A348354-10G
77115-084EA 15.92 USD
77115-084 77115-086 77115-088 77115-090 77612-982
5-Bromo-3-chloro-2-methoxypyridine 98%
5-Bromo-3-chloro-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClNO
MW: 222.47 g/mol
MDL Number: MFCD04972382
CAS Number: 848366-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR