2-Acetamidot-4-methylhiazole 95%

Supplier: Ambeed
Warning

Synonyms: N-(4-Methylthiazol-2-yl)acetamide

A344089-1G A344089-5G
77115-062EA 43.5 USD
77115-062 77115-064
2-Acetamidot-4-methylhiazole 95%
2-Acetamidot-4-methylhiazole

Precaución: for Research Use Only

Formula: C₆H₈N₂OS
MW: 156.2055 g/mol
MDL Number: MFCD00011561
CAS Number: 7336-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR