H-Phe(4-tBu)-OH 95%

Supplier: Ambeed
Warning

A342375-10G A342375-1G A342375-250MG A342375-5G
77115-040EA 196.73 USD
77115-040 77115-042 77115-044 77115-046
H-Phe(4-tBu)-OH 95%
H-Phe(4-tBu)-OH

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₂
MDL Number: MFCD02259528
CAS Number: 82372-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR