1,3,4,6-Tetrakis(methoxymethyl)glycoluril 97%

Supplier: Ambeed
Warning

Synonyms: 1,3,4,6-Tetrakis(methoxymethyl)tetrahydroimidazo[4,5-d]imidazole-2,5(1H,3H)-dione

A342019-100G A342019-25G
77115-030EA 19.57 USD
77115-030 77115-032
1,3,4,6-Tetrakis(methoxymethyl)glycoluril 97%
1,3,4,6-Tetrakis(methoxymethyl)glycoluril

Precaución: for Research Use Only

Formula: C₁₂H₂₂N₄O₆
MDL Number: MFCD00402438
CAS Number: 17464-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR