Tris(2-methylphenyl)phosphine 98%

Supplier: Ambeed
Warning

Synonyms: Tris(o-tolyl)phosphine, Tri-o-tolylphosphine

A341207-1000G A341207-100G A341207-10G A341207-250MG A341207-25G A341207-500G A341207-5G
77115-008EA 394.66 USD
77115-008 77115-010 77115-012 77115-014 77115-016 77115-018 77115-020
Tris(2-methylphenyl)phosphine 98%
Tris(2-methylphenyl)phosphine

Precaución: for Research Use Only

Formula: C₂₁H₂₁P
MW: 304.37 g/mol
MDL Number: MFCD00008514
CAS Number: 6163-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR