2-Amino-5-bromo-3-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-bromo-3-picoline, 5-Bromo-3-methylpyridin-2-amine, 5-Bromo-3-methyl-2-pyridylamine

A339583-100G A339583-10G A339583-1G A339583-25G A339583-500G A339583-5G
77114-992EA 56.64 USD
77114-992 77114-994 77114-996 77114-998 77115-000 77115-002
2-Amino-5-bromo-3-methylpyridine 98%
2-Amino-5-bromo-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MW: 187.04 g/mol
MDL Number: MFCD00068232
CAS Number: 3430-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR