Boc-Glu(OBzl)-OMe 95%

Supplier: Ambeed
Warning

Synonyms: Boc-Glu(Obzl)-OMe

A336677-10G A336677-1G A336677-5G
77114-968EA 76.22 USD
77114-968 77114-970 77114-972
Boc-Glu(OBzl)-OMe 95%
Boc-Glu(OBzl)-OMe

Precaución: for Research Use Only

Formula: C₁₈H₂₅NO₆
MDL Number: MFCD00190792
CAS Number: 59279-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR