Ciclopirox 97%

Supplier: Ambeed
Warning

A333509-100MG A333509-10G A333509-50MG A333509-5G A333509-1G A333509-1MG A333509-25G
77114-940EA 9.27 USD
77114-940 77114-942 77114-944 77114-946 77616-468 77685-952 77685-953
Ciclopirox 97%
Ciclopirox

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₂
MDL Number: MFCD00599441
CAS Number: 29342-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR