SB-3CT 98%

Supplier: Ambeed
Warning

A332833-10MG A332833-1MG A332833-50MG A332833-5MG
77114-924EA 30.9 USD
77114-924 77114-926 77114-928 77114-930
SB-3CT 98%
SB-3CT

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃S₂
MDL Number: MFCD09836266
CAS Number: 292605-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR