Methyl-2-oxocyclopentanecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-cyclopentanonecarboxylate

A332699-1000G A332699-100G A332699-10G A332699-500G A332699-5G A332699-25G
77114-914EA 74.16 USD
77114-914 77114-916 77114-918 77114-920 77114-922 77306-436
Methyl-2-oxocyclopentanecarboxylate 97%
Methyl-2-oxocyclopentanecarboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀O₃
MDL Number: MFCD00001411
CAS Number: 10472-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR