6,7-Dimethoxycoumarin 95%

Supplier: Ambeed
Danger

Synonyms: 6,7-Dimethoxy-2H-chromen-2-one

A332142-100MG A332142-10MG A332142-1G A332142-1MG A332142-250MG A332142-25MG A332142-50MG A332142-5G A332142-5MG
77114-896EA 49.87 USD
77114-896 77114-898 77114-900 77114-902 77114-904 77114-906 77114-908 77114-910 77114-912
6,7-Dimethoxycoumarin 95%
6,7-Dimethoxycoumarin

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₄
MW: 206.195 g/mol
MDL Number: MFCD00006871
CAS Number: 120-08-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR