3-Chlorobenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (3-Chlorophenyl)methanol

A331554-100G A331554-10G A331554-25G A331554-5G
77114-888EA 53.05 USD
77114-888 77114-890 77114-892 77114-894
3-Chlorobenzyl alcohol 98%
3-Chlorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₇ClO
MW: 142.58 g/mol
MDL Number: MFCD00004632
CAS Number: 873-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR