(3-Chloro-4-(methylthio)phenyl)boronic acid 98+%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-(methylthio)phenylboronic acid

A330305-100MG A330305-1G A330305-250MG A330305-5G
77114-866EA 38.12 USD
77114-866 77114-868 77114-870 77114-872
(3-Chloro-4-(methylthio)phenyl)boronic acid 98+%
(3-Chloro-4-(methylthio)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₈BClO₂S
MDL Number: MFCD22543649
CAS Number: 877383-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR