4-Fluoro-o-phenylenediamine 97%

Supplier: Ambeed
Warning

Synonyms: 3,4-Diamino-1-fluorobenzene, 4-Fluorobenzene-1,2-diamine, 1,2-Diamino-4-fluorobenzene, 4-Fluoro-1,2-phenylenediamine

A328448-10G A328448-25G A328448-1G A328448-5G
77114-848EA 39.15 USD
77114-848 77114-850 77571-942 77685-940
4-Fluoro-o-phenylenediamine 97%
4-Fluoro-o-phenylenediamine

Precaución: for Research Use Only

Formula: C₆H₇FN₂
MDL Number: MFCD00042228
CAS Number: 367-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR