5,6-Difluoroindoline-2,3-dione 97%

Supplier: Ambeed
Warning

Synonyms: 5,6-Difluoroisatin

A326888-100G A326888-1G A326888-5G
77114-822EA 879.49 USD
77114-822 77114-824 77612-884
5,6-Difluoroindoline-2,3-dione 97%
5,6-Difluoroindoline-2,3-dione

Precaución: for Research Use Only

Formula: C₈H₃F₂NO₂
MW: 183.110 g/mol
MDL Number: MFCD02998377
CAS Number: 774-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR