2,3,4,5-Tetramethoxytoluene 97%

Supplier: Ambeed
Warning

A326499-5G A326499-25G
77114-816EA 6.37 USD
77114-816 77114-818 77685-935
2,3,4,5-Tetramethoxytoluene 97%
2,3,4,5-Tetramethoxytoluene

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₄
MDL Number: MFCD07370118
CAS Number: 35896-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR