7-(2-Hydroxypropyl)theophylline ≥99%

Supplier: Ambeed
Warning

A325694-100MG A325694-5G A325694-25G A325694-1G
77114-808EA 6.18 USD
77114-808 77114-810 77612-876 77685-933
7-(2-Hydroxypropyl)theophylline ≥99%
7-(2-Hydroxypropyl)theophylline

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₄O₃
MDL Number: MFCD00022833
CAS Number: 603-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR