2-Methyl-5-nitrobenzenesulfonamide 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Aminosulphonyl)-4-methylnitrobenzene, 4-Nitro-2-sulphamoyltoluene

A325580-100G A325580-10G A325580-25G A325580-500G A325580-5G
77114-792EA 78.51 USD
77114-792 77114-794 77114-796 77114-798 77114-800
2-Methyl-5-nitrobenzenesulfonamide 97%
2-Methyl-5-nitrobenzenesulfonamide

Precaución: for Research Use Only

Formula: C₇H₈N₂O₄S
MW: 216.214 g/mol
MDL Number: MFCD00115503
CAS Number: 6269-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR