2,4-Diamino-6-ethoxypyrimidine 97%

Supplier: Ambeed
Warning

A325552-1G A325552-250MG
77114-788EA 61.8 USD
77114-788 77114-790
2,4-Diamino-6-ethoxypyrimidine 97%
2,4-Diamino-6-ethoxypyrimidine

Precaución: for Research Use Only

Formula: C₆H₁₀N₄O
MDL Number: MFCD09031077
CAS Number: 116436-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR