2-Bromo-5-(trifluoromethyl)benzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

A325439-1G A325439-5G
77114-784EA 17.51 USD
77114-784 77114-786
2-Bromo-5-(trifluoromethyl)benzenesulfonyl chloride 98%
2-Bromo-5-(trifluoromethyl)benzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₃BrClF₃O₂S
MDL Number: MFCD03094395
CAS Number: 176225-08-4
UN: 1760
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR