(R)-(+)-XylBINAP 98%

Supplier: Ambeed
Warning

Synonyms: (R)-(+)-2,2'-Bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl, (R)-2,2'-Bis(di-3,5-xylylphosphino)-1,1'-binaphthyl

A324365-100MG A324365-1G A324365-250MG A324365-5G
77114-764EA 11.34 USD
77114-764 77114-766 77114-768 77114-770
(R)-(+)-XylBINAP 98%
(R)-(+)-XylBINAP

Precaución: for Research Use Only

Formula: C₅₂H₄₈P₂
MDL Number: MFCD01630821
CAS Number: 137219-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR