3-Methoxy-4-carboxyphenylboronic acid 98%

Supplier: Ambeed
Warning

A324199-10G A324199-1G A324199-250MG A324199-5G
77114-756EA 323.58 USD
77114-756 77114-758 77114-760 77114-762
3-Methoxy-4-carboxyphenylboronic acid 98%
3-Methoxy-4-carboxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₅
MDL Number: MFCD06656278
CAS Number: 851335-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR