Fmoc-O-ethyl-D-tyrosine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-D-Tyr(4-Et)-OH, Fmoc-D-Tyr(Et)-OH
A323052-10G
A323052-1G
A323052-25G
A323052-5G
77114-732EA
31.94
USD
77114-732
77114-734
77114-736
77114-738
Fmoc-O-ethyl-D-tyrosine 97%
Fmoc-O-ethyl-D-tyrosine
Precaución: for Research Use Only
Formula:
C₂₆H₂₅NO₅ |
MDL Number:
MFCD00237035 CAS Number: 162502-65-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...