1-Chloro-2-phenylethane 98%

Supplier: Ambeed
Warning

Synonyms: 2-Phenylethyl chloride, (2-Chloroethyl)benzene, 2-(Chloroethyl)benzene

A317865-100G A317865-25G A317865-500G A317865-5G
77114-682EA 19.1 USD
77114-682 77114-684 77114-686 77114-688
1-Chloro-2-phenylethane 98%
1-Chloro-2-phenylethane

Precaución: for Research Use Only

Formula: ClCH₂CH₂C₆H₅
MW: 140.61 g/mol
MDL Number: MFCD00000977
CAS Number: 622-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR