5-Iodo-m-xylene 98%

Supplier: Ambeed
Warning

A312877-1000G A312877-100G A312877-10G A312877-25G A312877-500G A312877-5G
77114-646EA 295.61 USD
77114-646 77114-648 77114-650 77114-652 77114-654 77114-656
5-Iodo-m-xylene 98%
5-Iodo-m-xylene

Precaución: for Research Use Only

Formula: C₈H₉I
MDL Number: MFCD00060659
CAS Number: 22445-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR