2,3-Dichloro-5-methoxypyridine 98%

Supplier: Ambeed
Warning

A312076-100MG A312076-250MG A312076-5G
77114-632EA 70.02 USD
77114-632 77114-634 77114-636
2,3-Dichloro-5-methoxypyridine 98%
2,3-Dichloro-5-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅Cl₂NO
MW: 178.016 g/mol
MDL Number: MFCD08277295
CAS Number: 885168-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR