1-Methyl-2-oxo-2,3-dihydro-1H-1,3-benzodiazole-5-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-2-oxo-2,3-dihydro-1H-benzo[d]imidazole-5-carboxylic acid

A311900-1G A311900-250MG
77114-628EA 116.7 USD
77114-628 77114-630
1-Methyl-2-oxo-2,3-dihydro-1H-1,3-benzodiazole-5-carboxylic acid 95%
1-Methyl-2-oxo-2,3-dihydro-1H-1,3-benzodiazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈N₂O₃
MW: 192.18 g/mol
MDL Number: MFCD11131632
CAS Number: 19950-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR