4-Bromo-5-fluoro-2-methoxypyridine 97%

Supplier: Ambeed
Warning

A311656-250MG A311656-1G
77114-626EA 120.95 USD
77114-626 77685-888
4-Bromo-5-fluoro-2-methoxypyridine 97%
4-Bromo-5-fluoro-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFNO
MW: 206.01 g/mol
MDL Number: MFCD05662409
CAS Number: 884495-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR