(S)-4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine-6-carboxylic acid hydrochloride 95%

Supplier: Ambeed
Warning

A311234-100MG A311234-1G A311234-250MG
77114-618EA 13.39 USD
77114-618 77114-620 77114-622
(S)-4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine-6-carboxylic acid hydrochloride 95%
(S)-4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine-6-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₀ClN₃O₂
MDL Number: MFCD11519402
CAS Number: 88980-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR