1,3-Dimethoxybenzene 98%

Supplier: Ambeed
Warning

Synonyms: Resorcinol dimethyl ether

A310250-1000G A310250-100G A310250-10G A310250-25G A310250-500G
77114-598EA 108.22 USD
77114-598 77114-600 77114-602 77114-604 77114-606
1,3-Dimethoxybenzene 98%
1,3-Dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
MDL Number: MFCD00008384
CAS Number: 151-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR