Gemcitabine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 2'-Deoxy-2',2'-difluorocytidine, dFdC, 4-Amino-1-((2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride

A309564-100MG A309564-10G A309564-1G A309564-1MG A309564-250MG A309564-25G A309564-25MG A309564-5G
77114-570EA 10.61 USD
77114-570 77114-572 77114-574 77114-576 77114-578 77114-580 77114-582 77114-584
Gemcitabine hydrochloride 97%
Gemcitabine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂ClF₂N₃O₄
MW: 299.6591 g/mol
MDL Number: MFCD01735988
CAS Number: 122111-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR